Order Entry
Italy
ContactUsLinkComponent
1-Hydroxy-1-cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Irritant
1-Hydroxy-1-cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
1-Hydroxy-1-cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    107.0 to 111.0 deg-C

Specification Test Results

Frequently Bought Together