9-Hexyl-3,6-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:628336-95-8
:
Formula:C₃₀H₄₃B₂NO₄
MDL Number:MFCD20528097
:503.30 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:155.0 to 159.0 deg-C
Specification Test Results
Frequently Bought Together