Order Entry
Italy
ContactUsLinkComponent
 
 
1-Hexyl-3-methylimidazolium iodide ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
undefined
 
1-Hexyl-3-methylimidazolium iodide ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Brown to Dark purple clear liquid
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

 

Frequently Bought Together