
(E)-2'-Hydroxy-3,4-dimethoxychalcone ≥98.0% (by HPLC)
TCIAH1598-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:113.0 to 117.0 deg-C
:
:284.31 g/mol
:GHS07
:79140-20-8
:C₁₇H₁₆O₄