
4-Hydroxypyrrolidin-2-one ≥95.0% (by GC)
Catalog # TCIAH1573-500MG
Supplier: TCI
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:119.0 to 123.0 deg-C
- Specific rotation [a]20/D:-1.0 to +1.0 deg (C=1, H2O)
Specification Test Results
About this item
:
:101.11 g/mol
:GHS07
CAS Number:25747-41-5
Formula:C₄H₇NO₂
MDL Number:MFCD01687254