Order Entry
Italy
ContactUsLinkComponent
4-Hexylthiophene-2-carboxaldehyde ≥97.0% (by GC)
4-Hexylthiophene-2-carboxaldehyde ≥97.0% (by GC)
Catalog # TCIAH1499-1G
Supplier:  TCI
CAS Number:  
undefined
4-Hexylthiophene-2-carboxaldehyde ≥97.0% (by GC)
Catalog # TCIAH1499-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :196.31 g/mol
CAS Number:222554-30-5
Formula:C₁₁H₁₆OS
MDL Number:MFCD18414651