
1-(4-Hydrazinobenzyl)-1H-1,2,4-triazole dihydrochloride ≥97.0% (by HPLC, titration analysis)
Catalog # TCIAH1467-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Argentmetric Titration):min. 97.0 %
Specification Test Results
About this item
:
:262.14 g/mol
:GHS07
CAS Number:212248-62-9
Formula:C₉H₁₁N₅·₂HCl
MDL Number:MFCD06795497