
(S)-10-hydroxycamptothecin ≥98.0% (by HPLC)
Catalog # TCIAH1463-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
About this item
:
:364.35 g/mol
UN:1544
:GHS06
Melting Pt:267°C
Storage Temperature:Freezer
ADR:6.1,III
CAS Number:19685-09-7
:64439-81-2
Formula:C₂₀H₁₆N₂O₅
MDL Number:MFCD02093100