
(S)-4-Hydroxypyrrolidine-2-one ≥98.0% (by GC)
Catalog # TCIAH1368-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:155.0 to 159.0 deg-C
- Specific rotation [a]20/D:-57.0 to -61.0 deg (C=1、H2O)
Specification Test Results
About this item
:
:101.105 g/mol
:GHS07
CAS Number:68108-18-9
Formula:C₄H₇NO₂
MDL Number:MFCD00273370