1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1515-14-6
:
Formula:C₄H₄F₆O
MDL Number:MFCD00039259
:182.07 g/mol
Boiling Pt:62°C
EINECS:216-155-4
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together