(+/-)-cis-6-Hydroxy-1-methylbicyclo[4.3.0]nonane-2,7-dione ≥98.0% (by GC)
Catalog # TCIAH1318-50MG
Supplier: TCI
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:99.0 to 103.0 deg-C
- Solubility in Chloroform:almost transparency
Specification Test Results
About this item
:
:182.22 g/mol
CAS Number:1192178-33-8
Formula:C₁₀H₁₄O₃