Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:918-21-8
:
Formula:C₆H₂F₁₂O₂
MDL Number:MFCD00427700
:334.06 g/mol
Boiling Pt:129°C
Melting Pt:26°C
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together