Order Entry
Italy
ContactUsLinkComponent
1,1,1,3,3,3-Hexafluoroisopropyl trifluoromethanesulphonate ≥97.0% (by GC)
Corrosive
1,1,1,3,3,3-Hexafluoroisopropyl trifluoromethanesulphonate ≥97.0% (by GC)
Catalog # TCIAH1276-5G
Supplier:  TCI
CAS Number:  
Corrosive
1,1,1,3,3,3-Hexafluoroisopropyl trifluoromethanesulphonate ≥97.0% (by GC)
Catalog # TCIAH1276-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :300.09 g/mol
UN:1760
ADR:8,II
 :GHS05
CAS Number:156241-41-7
Formula:C₄HF₉O₃S
MDL Number:MFCD02093340