3-Hydroxyxanthen-9-one ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3722-51-8
:
Formula:C₁₃H₈O₃
MDL Number:MFCD00604748
:212.20 g/mol
Melting Pt:243°C
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together