
2-[(S)-1-Hydroxyethyl]pyridine ≥97.0% (by GC, titration analysis)
TCIAH1263-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
- Specific rotation [a]20/D:-26.0 to -32.0 deg (C=1, Toluene)
:
:123.16 g/mol
:29°C
:GHS07
:Refrigerator
:59042-90-9
:C₇H₉NO
:MFCD06795465