5-Hexyl-1,3-dithiolo[4,5-d][1,3]dithiole-2-thione ≥98.0% (by HPLC)
Catalog # TCIAH1163-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:41.0 to 45.0 deg-C
Specification Test Results
About this item
:
:294.52 g/mol
CAS Number:202126-51-0
Formula:C₁₀H₁₄S₅