Order Entry
Italy
ContactUsLinkComponent
 
4-(4-Hydroxybutyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
 :  TCI
 
4-(4-Hydroxybutyl)thiomorpholine-1,1-dioxide ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    63.0 to 66.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

Frequently Bought Together