4,4'-Hexamethylenebis(1,1-dimethylsemicarbazide) ≥98.0% (by titrimetric analysis)
: TCI
:69938-76-7
:
:C₁₂H₂₈N₆O₂
:MFCD00089122
:288.40 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:144.0 to 148.0 deg-C
- Solubility in hot Water:almost transparency
Frequently Bought Together