
Hexadecyltrimethylammonium hydroxide 10% in water
Supplier: TCI
Certificates
About this item
CAS Number:505-86-2
:
Formula:C₁₉H₄₃NO
MDL Number:MFCD00041921
:301.56 g/mol
EINECS:208-022-4
UN:3267
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Concentration (Neutralization titration):10.0 to 11.0 w/w%
Specification Test Results
Frequently Bought Together