Order Entry
Italy
ContactUsLinkComponent
 
1-Hexadecyl-4-methylpyridinium chloride hydrate ≥98.0% (by titrimetric analysis)
  TCIAH0967-5G
 :  TCI
 :  
1-Hexadecyl-4-methylpyridinium chloride hydrate ≥98.0% (by titrimetric analysis)
  TCIAH0967-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Argentmetric Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    max. 8.0 %

 

 

 :
 :354.02 g/mol
 :13106-53-1
 :C₂₂H₄₀ClN
 :MFCD02093475