Order Entry
Italy
ContactUsLinkComponent
4-Methyl-1-acetoxycalixarene [mixture of [6] and [8]] ≥95.0% (by HPLC) (contains 5-10% Acetone)
Flammable
4-Methyl-1-acetoxycalixarene [mixture of [6] and [8]] ≥95.0% (by HPLC) (contains 5-10% Acetone)
Supplier:  TCI
Certificates
undefined
Flammable
4-Methyl-1-acetoxycalixarene [mixture of [6] and [8]] ≥95.0% (by HPLC) (contains 5-10% Acetone)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Gray to Brown powder to crystal
  • Purity (HPLC):
    min. 95.0 area%

Specification Test Results

Related Information

Frequently Bought Together