Order Entry
Italy
ContactUsLinkComponent
(S)-(+)-3-Hydroxy-2,2-dimethylcyclohexanone ≥95.0% (by GC)
(S)-(+)-3-Hydroxy-2,2-dimethylcyclohexanone ≥95.0% (by GC)
Catalog # TCIAH0887-100MG
Supplier:  TCI
CAS Number:  
(S)-(+)-3-Hydroxy-2,2-dimethylcyclohexanone ≥95.0% (by GC)
Catalog # TCIAH0887-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 95.0 %
  • Specific rotation [a]20/D:
    +21.0 to +25.0 deg (C=1, CHCl3)

Specification Test Results

About this item

 :
 :142.20 g/mol
CAS Number:87655-21-8
Formula:C₈H₁₄O₂
MDL Number:MFCD00671542