Order Entry
Italy
ContactUsLinkComponent
 
4-(1-Hydroxyethyl)biphenyl ≥98.0% (by GC)
  TCIAH0866-5G
 :  TCI
 :  
4-(1-Hydroxyethyl)biphenyl ≥98.0% (by GC)
  TCIAH0866-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    94.0 to 98.0 deg-C
  • Solubility in Toluene:
    almost transparency

 

 

 :
 :198.27 g/mol
 :222-629-1
 :3562-73-0
 :C₁₄H₁₄O
 :MFCD00016859