Order Entry
Italy
ContactUsLinkComponent
Heptylmagnesium bromide 21% ∼1 mol/L in THF
Flammable
Corrosive
Irritant
Health
Heptylmagnesium bromide 21% ∼1 mol/L in THF
Catalog # TCIAH0822-250G
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
Irritant
Health
Heptylmagnesium bromide 21% ∼1 mol/L in THF
Catalog # TCIAH0822-250G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Concentration (Neutralization titration):
    21.0 to 24.0 w/w%

Specification Test Results

About this item

 :
 :203.41 g/mol
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:13125-66-1
Formula:C₇H₁₅BrMg
MDL Number:MFCD00013517