Order Entry
Italy
ContactUsLinkComponent
 
 
3-Hydroxypropanesulphonic acid ∼80% ∼7.8 mol/L in water (contains varying amounts of 3,3'-Oxydipropanesulfonic Acid)
 :  TCI
 
 
3-Hydroxypropanesulphonic acid ∼80% ∼7.8 mol/L in water (contains varying amounts of 3,3'-Oxydipropanesulfonic Acid)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Concentration (Neutralization titration):
    min. 80.0 w/w%

 

Frequently Bought Together