Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Lithium-DL-2-hydroxybutyrate ≥98.0% (by titrimetric analysis)

25 g
Catalog # TCIAH0580-25G
Supplier:   TCI
Lithium-DL-2-hydroxybutyrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAH0580-25G
Supplier:  TCI
CAS Number:  381716-41-2
Lithium-DL-2-hydroxybutyrate ≥98.0% (by titrimetric analysis)

25 g

CAS Number:  381716-41-2
Price
125,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  DL-2-Hydroxybutyric Acid Lithium Salt
  • CAS Number:381716-41-2
  • Formula:C₄H₈O₃
  • MDL Number:MFCD00040991
  • Molecular Weight:110.04 g/mol
Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ion exchange titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

Specifications

  • Pk:
    25 g

Specifications