Order Entry
Italy
ContactUsLinkComponent
1,5-Hexadiyne ≥95.0% (by GC) stabilised
Flammable
Irritant
Health
1,5-Hexadiyne ≥95.0% (by GC) stabilised
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
Health
1,5-Hexadiyne ≥95.0% (by GC) stabilised
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Yellow to Green clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Frequently Bought Together