Order Entry
Italy
ContactUsLinkComponent
4-Hydroxy-4-methyl-2-pentanone ≥98.0% (by GC)
Flammable
Irritant
Health
4-Hydroxy-4-methyl-2-pentanone ≥98.0% (by GC)
Catalog # TCIAH0272-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
4-Hydroxy-4-methyl-2-pentanone ≥98.0% (by GC)
Catalog # TCIAH0272-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :116.16 g/mol
Merck Index:14,02964
Boiling Pt:168°C
Melting Pt:-44°C
Melting Pt:–44°C
Flash Pt:58°C
EINECS:204-626-7
UN:1148
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
CAS Number:123-42-2
Formula:(CH₃)₂C(OH)CH₂COCH₃
MDL Number:MFCD00004471