
5-(Hydroxymethyl)-2-furaldehyde ≥95.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:67-47-0
:
Formula:C₆H₆O₃
MDL Number:MFCD00003234
Merck Index:14,04832
Flash Pt:79°C
:Stabilised with water
:126.11 g/mol
EINECS:200-654-9
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Yellow to Amber to Dark purple clear liquid
- Purity (GC):min. 95.0 %
- Water:3.0 to 5.0 %
Specification Test Results
Frequently Bought Together