



Hexadecyltrimethylammonium hydroxide 25% in methanol
Supplier: TCI
Certificates
About this item
CAS Number:505-86-2
:
Formula:C₁₉H₄₃NO
MDL Number:MFCD00041921
:301.56 g/mol
Flash Pt:11°C
EINECS:208-022-4
UN:2924
ADR:3,III
:GHS02
:GHS05
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Neutralization titration):25.0 to 27.0 w/w%
- Water:max. 5.0 %
Specification Test Results
Frequently Bought Together