
Graphene, nanoplatelets aggregates, sub-micronparticles surface area 750 m²/g
Catalog # TCIAG0502-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Specific surface:710 to 790 m2/g
Specification Test Results
About this item
:
:GHS07
CAS Number:1034343-98-0
:7782-42-5
MDL Number:MFCD00144065