G2 Glycan ≥95.0% (by HPLC)
Catalog # TCIAG0487-1MG
Supplier: TCI
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 95.0 area%
Specification Test Results
About this item
:
:1641.50 g/mol
Storage Temperature:Freezer
CAS Number:71496-53-2
Formula:C₆₂H₁₀₄N₄O₄₆
MDL Number:MFCD01310893