Gabexate mesylate ≥97.0% (by total nitrogen basis)
TCIAG0464-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 94.0 area%
- Purity (with Total Nitrogen):min. 97.0 %
- Melting point:89.0 to 93.0 deg-C
:
:417.5 g/mol
:14,04320
:56974-61-9
:C₁₆H₂₃N₃O₄·CH₃SO₃H
:MFCD00210299