Gestrinone ≥98.0% (by titrimetric analysis)
Catalog # TCIAG0436-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:150.0 to 154.0 deg-C
- Specific rotation [a]20/D:+83.0 to +86.0 deg (C=0.4, EtOH)
Specification Test Results
About this item
:
:308.42 g/mol
Merck Index:14,04415
:GHS08
Storage Temperature:Refrigerator
CAS Number:16320-04-0
Formula:C₂₁H₂₄O₂
MDL Number:MFCD00865561