Galaxolide(1,3,4,6,7,8-hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran) ∼50% in diethyl phthalate
Supplier: TCI
Certificates
About this item
CAS Number:1222-05-5
:
Formula:C₁₈H₂₆O
MDL Number:MFCD00217003
:258.41 g/mol
EINECS:214-946-9
UN:3082
ADR:9,III
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Concentration (GC):47.0 to 53.0 w/w%
Specification Test Results
Frequently Bought Together