Order Entry
Italy
ContactUsLinkComponent
Galβ(1-3)GlcNAcβ(1-3)Gal-β(1-4)Glc-β-pNP ≥97.0% (by HPLC)
Galβ(1-3)GlcNAcβ(1-3)Gal-β(1-4)Glc-β-pNP ≥97.0% (by HPLC)
Catalog # TCIAG0348-5MG
Supplier:  TCI
CAS Number:  
Galβ(1-3)GlcNAcβ(1-3)Gal-β(1-4)Glc-β-pNP ≥97.0% (by HPLC)
Catalog # TCIAG0348-5MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%

Specification Test Results

About this item

 :
 :828.73 g/mol
Storage Temperature:Freezer
CAS Number:148705-09-3
Formula:C₃₂H₄₈N₂O₂₃