Galβ(1-3)GalNAc-ɑ-Thr ≥97.0% (by HPLC)
Catalog # TCIAG0340-5MG
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Specific rotation [a]20/D:+83.0 to +87.0 deg (C=1, H2O)
Specification Test Results
About this item
:
:484.46 g/mol
Storage Temperature:Freezer
CAS Number:60280-58-2
Formula:C₁₈H₃₂N₂O₁₃
MDL Number:MFCD02683396