Glycerol-1,2-carbonate ≥90.0% (by GC)
Catalog # TCIAG0279-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 90.0 %
Specification Test Results
About this item
:
:118.09 g/mol
Boiling Pt:160°C
Flash Pt:212°C
EINECS:213-235-0
CAS Number:931-40-8
Formula:C₄H₆O₄
MDL Number:MFCD00085561