Order Entry
Italy
ContactUsLinkComponent
Ethyl glyoxylate 47% in toluene
Flammable
Irritant
Health
Ethyl glyoxylate 47% in toluene
Supplier:  TCI
Certificates
Flammable
Irritant
Health
Ethyl glyoxylate 47% in toluene
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorlessclear liquid
  • Concentration (Oxime Formation):
    47.0 to 51.7 w/w%

Specification Test Results

Related Information

Frequently Bought Together