Order Entry
Italy
ContactUsLinkComponent
(E/Z)-Geranylacetone mixture E/Z (3:2) ≥96.0% (by GC)
 
(E/Z)-Geranylacetone mixture E/Z (3:2) ≥96.0% (by GC)
  TCIAG0236-25ML
 :  TCI
 :  
 
(E/Z)-Geranylacetone mixture E/Z (3:2) ≥96.0% (by GC)
  TCIAG0236-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Purity (GC):
    min. 96.0 % (total of isomers)

 

 

 :
 :194.32 g/mol
 :211-711-2
 :GHS07
 :689-67-8
 :(CH₃)₂C=CHCH₂CH₂C(CH₃)=CHCH₂CH₂C(O)CH₃
 :MFCD00008910