Order Entry
Italy
ContactUsLinkComponent
Trisodium glycyrrhizinate hydrate ≥80.0% (by HPLC)
Trisodium glycyrrhizinate hydrate ≥80.0% (by HPLC)
Catalog # TCIAG0217-25G
Supplier:  TCI
CAS Number:  
Trisodium glycyrrhizinate hydrate ≥80.0% (by HPLC)
Catalog # TCIAG0217-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (HPLC):
    min. 80.0 area%
  • Specific rotation [a]20/D:
    +62.0 to +68.0 deg (C=1.5, H2O) (calcd.on anh.substance)
  • Water:
    max. 15.0 %
  • Sodium:
    7.1 to 8.1% (as Na) (calcd. Anh. Sub.)

Specification Test Results

About this item

 :
 :888.89 g/mol
CAS Number:71277-78-6
Formula:C₄₂H₆₂O₁₆
MDL Number:MFCD00070472