Orange II ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:633-96-5
:
Formula:C₁₆H₁₁N₂NaO₄S
MDL Number:MFCD00011657
:350.32 g/mol
Merck Index:14,06858
EINECS:211-199-0
:GHS07
Specification Test Results
- Appearance:Orange to Brown to Dark red powder to crystal
- Purity (HPLC):min. 97.0 area%
- Lambda max.:482.0 to 486.0 nm (0.02mol/L AcONH4 sol.)
- Absorbance (E1%1cm):min. 500 (0.02mol/L AcONH4 sol., 482.0 to 486.0 nm)
Specification Test Results
Frequently Bought Together