Triolein >50% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:122-32-7
:
Formula:C₅₇H₁₀₄O₆
MDL Number:MFCD00137563
:885.45 g/mol
Merck Index:14,09732
Boiling Pt:240°C
Melting Pt:–4°C
EINECS:204-534-7
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (HPLC):min. 50.0 area%
Specification Test Results
Frequently Bought Together