Fmoc-N-Me-Ile-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:138775-22-1
:
Formula:C₂₂H₂₅NO₄
MDL Number:MFCD00153389
:367.445 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:172.0 to 177.0 deg-C
- Specific rotation [a]20/D:-59.0 to -64.0 deg (C=1,DMF)
Specification Test Results
Frequently Bought Together