
3-Fluoro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
: TCI
:505083-04-5
:
:C₈H₈BFO₄
:MFCD04115665
:197.96 g/mol
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):97.0 to 110.0 %
Frequently Bought Together