Order Entry
Italy
ContactUsLinkComponent
4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic acid ≥97.0% (by HPLC)
 
4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic acid ≥97.0% (by HPLC)
  TCIAF1050-200MG
 :  TCI
 :  
undefined
 
4-[(9H-Fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic acid ≥97.0% (by HPLC)
  TCIAF1050-200MG
 :  TCI
 :  

 

  • Pk:
    200 mg
  • Seq:
    2530

 

 

  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Neutralization titration):
    min. 95.0 %

 

 

 :
 :353.37 g/mol
 :GHS07
 :Refrigerator
 :204320-51-4
 :C₂₀H₁₉NO₅
 :MFCD01632020