
9-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-4,7-dioxanonanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:872679-70-4
:
Formula:C₂₂H₂₅NO₆
MDL Number:MFCD08064306
:399.44 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:95.0 to 99.0 deg-C
Specification Test Results
Frequently Bought Together