N-[(9H-Fluoren-9-ylmethoxy)carbonyl]anthranilic acid ≥96.0% (by HPLC, titration analysis)
Catalog # TCIAF0923-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Neutralization titration):min. 96.0 %
Specification Test Results
About this item
:
:359.38 g/mol
CAS Number:150256-42-1
Formula:C₂₂H₁₇NO₄
MDL Number:MFCD00235883