N-Fmoc-β-alanine ≥99.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:35737-10-1
:
Formula:C₁₈H₁₇NO₄
MDL Number:MFCD00063328
:311.3 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:146.0 to 150.0 deg-C
Specification Test Results
Frequently Bought Together