



Fluazinam ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF0869-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:116.0 to 120.0 deg-C
Specification Test Results
About this item
:
:465.09 g/mol
Merck Index:14,04117
UN:2811
ADR:6.1,II
:GHS05
:GHS07
:GHS08
:GHS09
CAS Number:79622-59-6
Formula:C₁₃H₄Cl₂F₆N₄O₄
MDL Number:MFCD00214168